missing translation for 'onlineSavingsMsg'
Learn More

Phosphate-P Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 600

Code : 04

Additional Details : Weight : 0.59000kg

Quantity 500mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 11440189

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
PO&subscript_4;-P1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

Phosphate-Phosphorus Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specifications

Phosphate-Phosphorus Standard
99.9
10002g/mL
With Certificate of Analysis
H2O
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
99.9
7732-18-5
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
Videos
Documentation

Product intended for laboratory use only