Learn More
Nitrite Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand:  SPEX Certiprep ORG-CA8-4Z
Code : 04
Additional Details : Weight : 0.09000kg
Quantity 125mL
Packaging Triple-leached LDPE bottle
 View more versions of this product
Product Code. 11470279
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
Product intended for laboratory use only
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
AnalyteConcentration
NO&subscript_2;&superscript_minus;1000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chemical Identifiers
Nitrite Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Specifications
Nitrite Standard
99.9
1000μg/mL
With Certificate of Analysis
H2O
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
99.9
7732-18-5
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
Videos