missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep AS-NO39-2X

Code : 04

Additional Details : CAS Number : 7757-79-1 Weight : 0.09000kg

Packaging Triple-leached LDPE bottle
Quantity 500mL

 View more versions of this product

Product Code. 10141572

Quantity Price
1 £93.18 / 500mL
10 £85.44 / 500mL
Estimated Shipment
17-12-2020

Please to purchase this item. Need a web account? Register with us today!



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
NO&subscript_3;&superscript_minus;1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7732-18-5

Specifications

Nitrate Standard
7732-18-5
99.9
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
99.9
1000μg/mL
With Certificate of Analysis.
H2O
Videos
SDS Documents

Product intended for laboratory use only