missing translation for 'onlineSavingsMsg'
Learn More

Iodophenpropit dihydrobromide, Tocris Bioscience™

Potent, selective H3 antagonist

Brand:  Tocris (Bio-Techne) 077910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13427993

  • £139.65 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective H3 antagonist with high affinity (KD = 0.3 nM). Also available as part of the Histamine H3 Receptor Tocriset™ .
Specifications

Chemical Identifiers

145196-87-8
576.133
iodophenpropit dihydrobromide, iodophenpropit hydrobromide, 3-1h-imidazol-4-yl propyl n-2-4-iodophenyl ethyl carbamimidothioate dihydrobromide, n-2-4-iodophenyl ethyl-s-3-4 5-imidazolyl propyl isothiourea dihydrobromide
3-(1H-imidazol-5-yl)propyl N'-[2-(4-iodophenyl)ethyl]carbamimidothioate;dihydrobromide
C15H21Br2IN4S
BOSOGNBLIWPCMS-UHFFFAOYSA-N
24978528
C1=CC(=CC=C1CCN=C(N)SCCCC2=CN=CN2)I.Br.Br

Specifications

10mg
145196-87-8
iodophenpropit dihydrobromide, iodophenpropit hydrobromide, 3-1h-imidazol-4-yl propyl n-2-4-iodophenyl ethyl carbamimidothioate dihydrobromide, n-2-4-iodophenyl ethyl-s-3-4 5-imidazolyl propyl isothiourea dihydrobromide
C1=CC(=CC=C1CCN=C(N)SCCCC2=CN=CN2)I.Br.Br
576.133
576.13
Iodophenpropit dihydrobromide
C15H21Br2IN4S
BOSOGNBLIWPCMS-UHFFFAOYSA-N
3-(1H-imidazol-5-yl)propyl N'-[2-(4-iodophenyl)ethyl]carbamimidothioate;dihydrobromide
24978528
>98%
Videos
Documents