Learn More
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Marca: Thermo Scientific Acros 378418000
Code : 29
Detalles adicionales : CAS Number : 74-88-4 Weight : 0.74640kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentEspecificaciones
Iodomethane | |
74-88-4,1634-04-4 | |
41.0°C to 43.0°C | |
AcroSeal™ Glass Bottle | |
MFCD00001073 | |
0.933 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CI | |
141.94 | |
CHEBI:39282 |
2M Solution in tert-Butyl Ether | |
0.9330g/mL | |
−18°C | |
CH3I | |
800 mL | |
15, 6159 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
6328 | |
141.94 |
RUO – Research Use Only