Learn More
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 378411000
Code : 29
Additional Details : CAS Number : 74-88-4 Weight : 0.09330kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Iodomethane | |
74-88-4,64-19-7 | |
41.0°C to 43.0°C | |
AcroSeal™ Glass bottle | |
MFCD00001073 | |
0.933 | |
mercury 2+ ion acetic acid | |
CC(=O)[O-].CC(=O)[O-].[Hg+2] | |
141.94 | |
CHEBI:33211 |
2M Solution in tert-Butyl Ether | |
0.9330g/mL | |
−18°C | |
CH3I | |
100 mL | |
15, 6159 | |
BRMYZIKAHFEUFJ-UHFFFAOYSA-L | |
mercury(2+);diacetate | |
15337 | |
141.94 |
RUO – Research Use Only