missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog Number p-7021912
Click to view available options
Quantity:
100 g
500 g
2500 g
75-03-6
C2H5I
155.966
MFCD00001091
HVTICUPFWKNHNG-UHFFFAOYSA-N
ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
6340
CHEBI:42487
iodoethane
CCI
75-03-6
C2H5I
155.966
MFCD00001091
HVTICUPFWKNHNG-UHFFFAOYSA-N
ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
6340
CHEBI:42487
iodoethane
CCI

Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
TRUSTED_SUSTAINABILITY
Melting Point -108°C
Density 1.95
Boiling Point 70°C to 73°C
Flash Point >73°C (163°F)
Refractive Index 1.512
Quantity 100 g
Beilstein 505934
Sensitivity Light and moisture sensitive
Merck Index 14,3813
Solubility Information Miscible with water.
Formula Weight 155.97
Percent Purity ≥98%
Chemical Name or Material Iodoethane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'