missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution 1% (w/v) In Potassium Iodide, Pure, Fisher Chemical™

Brand:  Fisher Chemical J/4450/17

Code : 23

Additional Details : Weight : 2.50000kg

Quantity 2.5L

 View more versions of this product

Product Code. 10122100

  • £86.89 / 2.50 L

In stock, ready to ship: 21

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Iodine Solution 1% (w/v) In Potassium Iodide, Pure

Specifications

Chemical Identifiers

11355

Specifications

11355
Amber glass bottle
2.5L
Liquid
Amber
Videos
SDS Documents
Certificates