missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, resublimed crystals, 99.9985% (metals basis), Thermo Scientific Chemicals, Puratronic™
Click to view available options
Quantity:
2.5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 183°C to 186° (sublimation) |
Color | Black-Blue |
Physical Form | Solid |
Quantity | 100 g |
Assay Percent Range | 99.9985% (Metals basis) |
IUPAC Name | molecular iodine |
Formula Weight | 253.81 |
Odor | Irritating |
Appearance | Bluish-black crystals |
Density | 4.93 g/cm3 at 20°C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'