missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, 1M solution in dichloromethane, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 370748000

Code : 29

Additional Details : CAS Number : 7790-99-0 Weight : 1.13600kg Transport : UN number : 2922 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 10582252

  • £219.00 / 800mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Iodine monochloride
7790-99-0,75-09-2
Solution
ClI
MFCD00011354
15, 5060
Solubility in water: insoluble
ClI
162.35
162.35
Glass bottle
1.4200g/mL
1, 0M solution in dichloromethane
Brown to Orange
800 mL
ICl
01,502; 08,261; 10,212; 11,268
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodochlorane
24640
0.95 to 1.05M
1.42
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iodine monochloride, 1,0M solution in dichloromethane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only