missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98% min, Thermo Scientific Chemicals

Catalog Number 11303488
Click to view available options
Quantity:
25 g
100 g
7789-33-5
BrI
206.81
MFCD00011353
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
82238
iodobromane
BrI
7789-33-5
BrI
206.81
MFCD00011353
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
82238
iodobromane
BrI

Iodine bromide is used for bromination of and for determining iodine used. IBr is used in some types of iodometry. It is also used as a organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 116°C (decomposition)
Melting Point 50° (sublimation)
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98% min
Beilstein 3902973
Flash Point None
Solubility Information It is soluble in alcohol,water,ether,carbon disulfi,and glacial acetic acid.
Formula Weight 206.81
Sensitivity Light and moisture sensitive
Density 4.416
Chemical Name or Material Iodine bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'