missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution

Brand:  Thermo Scientific Chemicals 035636.K7

Code : NEW

Additional Details : CAS Number : 7553-56-2 Weight : 0.01000kg

 View more versions of this product

Product Code. 11303767

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Iodine, 0.01N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iodine
4 L
I2
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.81
23 hPa (17mm Hg) at 20°C
Liquid
7553-56-2,7681-11-0,7732-18-5
Red to Brown
MFCD00011355 MFCD00164163
II
253.81
CHEBI:17606
Iodine: 0.13%; Potassium iodide: 2.5%; Water: 97.37%
Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only