missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, 99.5%, for analysis, ACROS Organics™

Brand:  Acros Organics 206452500

Code : 29

Additional Details : CAS Number : 7782-68-5 Weight : 0.30000kg Transport : UN number : 3085 Chem class : 1a Pack group : II

Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10409910

  • £129.10 / 250g


Estimated Shipment
14-03-2018
Add to basket

Specifications

Chemical Identifiers

Iodic acid
7782-68-5
99.5%
HIO3
175.91
p.a.
For analysis
99.5%
HIO3
MFCD00011348

Specifications

Iodic acid
250g
7782-68-5
Crystalline Powder
99.5%
HIO3
MFCD00011348
15, 5056
10ppm max.
5ppm max.
Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform
100ppm max.
White to light beige
p.a.
For analysis
99.5%
HIO3
175.91
0.01% max. (as Cl)
4.6200g/mL
10ppm max.
Glass bottle
4.62
110.0°C
Videos
SDS