Learn More
Invitrogen™ TOPO™ XL PCR Cloning Kit, with One Shot™ TOP10 Chemically Competent E. coli
Combines a unique vector and novel protocol for easy, efficient cloning of long (3-10kb) PCR products
Brand:  Invitrogen™ K475010
Code : 50
Additional Details : Weight : 0.38000kg
 View more versions of this product
Product Code. 10575193
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Kit, PCR Cloning
Each TOPO XL PCR Cloning Kit includes linearized and topoisomerase I-activated pCR-XL-TOPO vector, dNTPs, control template and primers, M13 forward and reverse primers, sodium iodide solution, binding buffer, 4X final wash, S.N.A.P. purification columns and collection vials, 6X crystal violet loading buffer, 2mg/mL crystal violet solution, One Shot Electrocomp or Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid
10 reactions
TOP10
Cloning, PCR Cloning
Videos