Learn More
Invitrogen™ ThermoScript™ RT-PCR System for First-Strand cDNA Synthesis
Brand: Invitrogen™ 11146016
Code : 50
Additional Details : Weight : 0.00025kg
Specifications
100μL ThermoScript RT (15U/μL); 500μL 5X cDNA Synthesis Buffer; 250μL 0.1M DTT; 2 × 250μL 10mM dNTP Mix; 100μL RNaseOUT (40U/μL); 100μL Oligo (dT)20 (50μM); 250μL Random Hexamers (50ng/μL); 1.25mL DEPC-Treated Water; 2 × 50μL E. coli Rnase H (2U/μL). Store at -20deg.C | |
Random Primers, Oligo dT Primers | |
100 reactions | |
cDNA Synthesis, RT-PCR |