missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T4 Polynucleotide Kinase

Brand:  Invitrogen 18004010

Code : 50

Additional Details : Weight : 0.00100kg

Product Code. 10790004

  • £92.50 / Each


Estimated Shipment
04-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Specifications

Specifications

A vial of 5X forward reaction buffer [350mM Tris-HCl (pH 7.6), 50mM MgCl2 , 500mM KCl, 5mM 2-mercaptoethanol] and a vial of 5X exchange reaction buffer [250mM imidazole-HCl (pH 6.4), 60mM MgCl2 , 5mM 2-mercaptoethanol, 350μM ADP].
200U
Store at -20°C
Videos