Learn More
Invitrogen™ Quant-iT™ Qubit™ dsDNA BR Assay Kit
Highly selective for double-stranded DNA (dsDNA) over RNA
£66.25 - £195.00
Requires: Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10625443 Invitrogen™
Q32850
100 assay £66.25
1 set View alternative products
Save Up to 
»
EU Stock 188 Please to purchase this item. Need a web account? Register with us today!
10146592 Invitrogen™
Q32853
500 assay £195.00
500 assays View alternative products
Save Up to 
»
EU Stock 193 Please to purchase this item. Need a web account? Register with us today!
Description
Description
The Qubit dsDNA BR Assay Kit is designed specifically for use with the Qubit Fluorometer. The assay is highly selective for double-stranded DNA (dsDNA) over RNA and is designed to be accurate for initial sample concentration from 100 pg/µl–1,000 ng/µl. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µl and 20 µl is acceptable), and read the concentration using the Qubit Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.
Which product to choose for dsDNA BR (Broad Range) quantitation?
  • For 1–20 samples: use this Qubit dsDNA BR Assay Kit with the Qubit Fluorometer
  • For 20–2000 samples: use the Quant-iT dsDNA BR Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 µL thin-walled PCR tubes are required but not included.
Order Info
Shipping Condition: Room temperature
For Research Use Only. Not for use in diagnostic procedures.