missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix Universal

Incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms including ABI 7500, Corbett Rotor-Gene™ etc.

Brand:  Invitrogen™ 1176202K

Code : 50

Additional Details : Weight : 4.00000kg

 View more versions of this product

Product Code. 10543533

  • £2816.28 / Each


EU Stock 2
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range

Consistent Specificity and Low-Copy Detection:
  • The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets
  • The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity
  • With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA
Specifications

Specifications

Chromatin biology, real time PCR (qPCR), real time PCR-based gene expression profiling
2000 Reactions
Detection Method: SYBR; PCR Method: qPCR
DNA (Genomic), cDNA
2X SYBR™ GreenER™ qPCR SuperMix and a separate tube of ROX reference dye.
Store at 4°C upon receipt.
7500 System, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Standard
Videos