missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument

Incorporate the latest high-performance technology to provide the most reliable gene expression data from your ABI 7300, 7000, 7700 or 7900HT instrument.

£262.00 - £3910.00

Specifications

Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Product Line Platinum™, SYBR™ GreenER™
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10741024
View Documents Promotion Details
Invitrogen™
1176002K
2000 Reactions
£3910.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10656413
View Documents Promotion Details
Invitrogen™
11760500
500 Reactions
£1136.00
500 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10512773
View Documents Promotion Details
Invitrogen™
11760100
100 Reactions
£262.00
100 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.
Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

  • The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
  • A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
  • An optimized buffer system - improves sensitivity and provides excellent long-term stability
  • Uracil DNA glycosylase - reduces carryover contamination in qPCR

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

SYBR
qPCR
Platinum™, SYBR™ GreenER™
Gene Expression
DNA (Genomic), cDNA
High
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Real Time PCR SYBR Master Mix
2X
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.