Learn More
Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG
Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.
Brand: Invitrogen™ 11733038
178.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.04454kg
Description
The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.
- Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
- 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
- Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
- SYBR™ Green I dye for simple and easy detection of real-time product
Order Info
Shipping Conditions: Dry Ice
Specifications
• Platinum™ SYBR™ Green qPCR SuperMix-UDG (2 × 1.25 mL) • 50 mM Magnesium Chloride (1 mL) • ROX Reference Dye (100 μL) • 20X BSA (300 μL) Sufficient reagents are provided for 100 reactions of 50 μL each. Store all components at -20°C. |
|
Tube | |
qPCR | |
Standard | |
Platinum™, SYBR™ | |
Dry Ice | |
Gene Expression | |
DNA (Genomic), cDNA |
SYBR | |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ | |
Real Time PCR SYBR Master Mix | |
100 Reactions of 50 μL Each | |
100 Reactions | |
2 x 1.25 mL |
For Research Use Only. Not for use in diagnostic procedures.