missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, high fidelity

Platinum™ Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.

Brand:  Invitrogen™ 11304029

Code : 50

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 10665783

  • £1100.00 / 500 reactions
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Platinum™ Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum™ Taq DNA Polymerase and the proofreading (3´→5´ exonuclease activity) enzyme Pyrococcus species GB-D polymerase. PCR specificity is improved with the incorporation of Platinum™ automatic 'hot-start' technology. Features of Platinum™ Taq DNA Polymerase, High Fidelity:

Fidelity—greater than six times higher fidelity than Taq DNA polymerase
Amplicon size—amplification of fragments up to 15 kb (see figure)
Convenience—room temperature reaction assembly
Applications—amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR

Unit definition
One unit of Platinum™ Taq DNA Polymerase, High Fidelity, is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 min at 74°C.

Specifications

Specifications

• Platinum Taq DNA Polymerase High Fidelity (1 x 100 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 2.25 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.
Low
Standard
Dry Ice
6X
500 Reactions
Separate Components
Primer-probe
Platinum Taq DNA Polymerase High Fidelity
DNA Polymerase High Fidelity
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Mixed
20 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.