PCR SuperMix

Ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification

Overview
Brand: Invitrogen

Manufacturer Part Number: 10572063

5000 RXN PCR SUPERMIX 5000 REACTIONS 00 STORE AT-20°C

UNSPSC: 12352204

Code: 50

Additional Details:
Additional Details: Weight: 0.04000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10514093

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Polymerase Taq DNA Polymerase
Size (Final Product) 5 kb or less
GC-Rich PCR Performance Low
Overhang 3'-A
Format SuperMix
Fidelity 1 X
Quantity 5000 reactions
Product Type PCR SuperMix
Exonuclease Activity 5' - 3'
For Use With (Application) Efficient PCR amplifications
Includes Contains 4 bottles (56.25mL each) of PCR SuperMix. Components: 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM Magnesium Chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, 22U/mL recombinant Taq DNA Polymerase, and stabilizers
No. of Reactions 5000.0