missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCR Optimized Buffers

Use with PCR Optimizer Kit to provide optimal buffer conditions for all primer-template combinations

Brand:  Invitrogen™ K122002G

UNSPSC : 41106307

Code : 50

Additional Details : Weight : 0.03000kg

 View more versions of this product

Product Code. 10338192

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Specifications

The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
Taq DNA Polymerase
Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
200.0
Buffer G