Invitrogen™ PCR Optimized Buffers

Use with PCR Optimizer Kit to provide optimal buffer conditions for all primer-template combinations

Overview
Brand: Invitrogen™

Manufacturer Part Number: K122002F

KIT,PCR OPTIMIZER BUF.FThe PCR Optimizer Kit is designed to give the

UNSPSC: 41106307

Code: 50

Additional Details:
Additional Details: Weight: 0.05000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10145032

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Polymerase Taq DNA Polymerase
Storage Requirements Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Product Type Buffer F
Includes The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
No. of Reactions 200.0