Learn More
Invitrogen™ iBlot™ Electrode Replacement Kit
For use with the iBlot Gel Transfer Device
Brand: Invitrogen™ IB1002
Code : 50
Additional Details : Weight : 0.00450kg
Includes: Two electrical contacts
Description
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
iBlot | |
Electrode Replacement Kit |
For Research Use Only. Not for use in diagnostic procedures.