Learn More
Pfx50™ DNA Polymerase
Designer enzyme ideal for highest fidelity and initial PCR success
Brand:  Invitrogen 12355036
Code : 50
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10073462
  • £420.00 / 500 reactions
Estimated Shipment
08-02-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Fidelity (vs. Taq): 50X
Pfx50 DNA Polymerase is supplied with a vial of 10X Pfx50 PCR amplification buffer contains 1.2mM MgSO4 and a vial of 50mM MgSO4 for further optimization.
Store at -20°C. Guaranteed stable for 6 months when properly stored.
Gel Compatibility: Low
500 reactions
Videos
Documentation
Technical Documents