Learn More
Invitrogen™ Pfx50™ DNA Polymerase
Brand: Invitrogen 12355012
Code : 50
Additional Details : Weight : 0.00454kg
Specifications
Pfx50 DNA Polymerase is supplied with a vial of 10X Pfx50 PCR amplification buffer contains 1.2mM MgSO4 and a vial of 50mM MgSO4 for further optimization. Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
Gel Compatibility: Low | |
100 reactions | |
Fidelity (vs. Taq): 50X |