Learn More
Invitrogen™ GeneCatcher™ gDNA Blood Kit, 3-10 mL
High sensitivity for archived or poorly stored blood samples
Brand: Invitrogen™ CS21110
Code : 50
Additional Details : Weight : 0.05250kg
Includes: GeneCatcher Magnetic Beads, Protease, Protease Buffer, GeneCatcher Lysis and Wash Buffers, and GeneCatcher Elution Buffer.
Description
ChargeSwitch™ gDNA 3-10 ml Blood Kit uses magnetic bead-based technology to allow rapid and efficient extraction of genomic DNA (gDNA) from human blood, including archived or poorly stored blood samples, without the use of centrifugation
- Rapid, three-step protocol
- Validated for automation
- Ideal for any downstream application of choice
- User-friendly protocol
- Flexible solution for large scale genotyping and biobanking projects
- Improved performance in downstream applications
DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Blood (Large-Scale, >1mL)
Order Info
Shipping Condition: Room Temperature
Specifications
The GeneCatcher™ gDNA kits include GeneCatcher™ Magnetic Beads, Protease, Protease Buffer, GeneCatcher™ Lysis and Wash Buffers, and GeneCatcher™ Elution Buffer. All components of the GeneCatcher™ gDNA Blood Kits are shipped at room temperature. Upon receipt, store the protease at +4°C and the remaining kit components at room temperature. All components are guaranteed stable for 6 months when properly stored. | |
Blood | |
Fewer resources used and less waste, less hazardous | |
> 1 ml Blood Samples (Large-Scale) | |
Room Temperature |
Magnetic Bead | |
Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, Forensic Testing | |
Not High-throughput Compatible (Manual) | |
GeneCatcher™ | |
Genomic DNA |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.