Learn More
Invitrogen™ Electrocomp™ TOP10F Electrocomp Kit
Identical to TOP10 with the addition of an F' episome
Brand:  Invitrogen™ C66524
Code : 50
Additional Details : Weight : 0.12000kg
Product Code. 10163762
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Bacterial/Yeast Strain: TOP10F
Cloning, Transformation
Specifications
Specifications
Contains enough electrocompetent cells for 75 x 40μL rxns or 50 x 20μL rxns
30 x 100 μL