Learn More
Invitrogen™ E-Gel™ SizeSelect Agarose Gels, 2%
For the convenient purification of DNA libraries for Next Generation Sequencing applications
Brand: Invitrogen™ G661002
Code : 50
Additional Details : CAS Number : 79-06-1 Weight : 0.01000kg
Includes: 10 E-Gel SizeSelect 2% Agarose
Description
- Separate and recover DNA for library construction in three steps: load, run, and retrieve
- Avoid additional gel purification steps required by other methods
- Remove purified DNA directly from the well with a pipette
- Minimize DNA damage by using blue-light transillumination
- Eliminate tedious and error-prone gel extraction steps
- Features two rows of wells: To extract a DNA band of interest without conventional gel purification, load the sample into the top row and electrophorese until the band moves into the bottom row of wells
- Then use a pipette to remove the purified DNA in the appropriate size range
- Guides on the gel cassette help track the target DNA band
- The direct access to the sample from the retrieval well allows the user to avoid the need to excise gel band slices and risk losing DNA during a gel purification step
- Gels run in the E-Gel™ iBase™ Power System: This self-contained device includes a built-in power supply
- View program selection and running time on the easy-to-read LCD display
- Preset programs are available for various gel types or manually set run times
- A reverse program allows the user to change the field direction, ensuring complete capture of DNA
- Automatic shutoff feature
- The E-Gel iBase™ Power System fits on a Safe Imager™ Blue-Light Transilluminator, to safely monitor DNA bands migrate in real-time: Blue-light transillumination can be used to view the proprietary stain
- Sample Type: Double-Stranded DNA (dsDNA)
- Sample Volume; 25μL
SOLiD Next-Generation Sequencing, SOLiD Next-Generation Sequencing Library Construction, Sample and Library Preparation for SOLiD Next-Generation Sequencing, Sequencing
Order Info
Shipping Condition: Room Temperature
Specifications
2% | |
Agarose Gel Cassette | |
2 x 8-well | |
10 Pack |
Multiwell | |
50 to 1000 bp | |
10 E-Gel SizeSelect 2% Agarose | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.