Learn More
Invitrogen™ Dynabeads™ Untouched™ Mouse CD4 Cells Kit
Dynabeads™ Untouched™ Mouse CD4 Cells contains both magnetic beads and an Antibody Mix to deplete all leucocytes from mouse spleen or lymph node cells, leaving only the target mouse CD4+ cells.
£226.00 - £768.00
Specifications
Cell Type | T Cells (CD4) |
---|---|
Product Type | Cell Isolation Kit |
Isolation Technology | Negative Isolation |
Product Line | DYNAL™, Dynabeads™ |
Purity or Quality Grade | Research Grade |
Includes: 20mL Mouse Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process approx. 1 x 109 cells
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10236794
|
Invitrogen™
11416D |
4 mL |
£226.00
4mL |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
10252133
|
Invitrogen™
11415D |
2 x 10 mL |
£768.00
Pack of 2 |
Estimated Shipment: 08-04-2024
Log in to see stock. |
|||||
Description
Dynabeads Untouched Mouse CD4 Cells supplies both 4.5μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix, together designed for the depletion of CD4-negative leucocytes from mouse spleen or lymph node single-cell suspensions. Following the depletion procedure, the negatively isolated, untouched target mouse CD4+ cells are pure, viable, and ready to be used in any downstream application.
- Rapid isolation of untouched mouse CD4
- T cells—no columns required
- Pure and viable mouse CD4+ T cells are ready for any downstream application
Order Info
Shipping Condition: Room Temperature
Specifications
T Cells (CD4) | |
Negative Isolation | |
Research Grade | |
Room Temperature | |
4.5 μm |
Cell Isolation Kit | |
DYNAL™, Dynabeads™ | |
Mouse | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.