Learn More
Invitrogen™ ChargeSwitch™ NoSpin Plasmid Micro Kit
Optimized for automation
Brand:  Invitrogen™ CS1020110
Code : 50
Additional Details : Weight : 1.29000kg
 View more versions of this product
Product Code. 10313143
  • £1067.15 / Each
Estimated Shipment
10-07-2018
Add to basket
Requires : Magnetic accessory
Specifications
Specifications
Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
960 preps
Bacterial Culture
Plasmid DNA
ChargeSwitch™ Chemistry
>100 μg (Small-Scale) Plasmid DNA
Store as follows: ChargeSwitch™ Precipitation Buffer (N5) at 4°C. Mix the RNase I in ChargeSwitch™ Resuspension Buffer (R4) and store the resulting solution at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.
Components:
ChargeSwitch™ MagnaClear™ Beads
ChargeSwitch™ Magnetic Beads
ChargeSwitch™ Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
ChargeSwitch™ Lysis Buffer (L9)
ChargeSwitch™ Precipitation Buffer (N5)
ChargeSwitch™ Wash Buffer (W11)
ChargeSwitch™ Wash Buffer (W12)
ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
Videos
Documentation
Technical Documents
For Research Use Only. Not for use in diagnostic procedures.