Learn More
Invitrogen™ RNA Century™-Plus Marker Templates
Contain mixtures of linearized plasmids ready for use as templates during in vitro transcription reactions for synthesis of labeled RNA molecular size standards
Brand: Invitrogen™ AM7782
Code : 50
Additional Details : Weight : 0.10500kg
Description
When transcribed with T7 RNA polymerase, the templates produce 7 RNA transcripts with lengths of 100, 200, 300, 400, 500, 750, and 1000 bases. They can be labeled with radioisotopes, biotin, or other nonisotopic modified nucleotides. They are ideal as size standards for ribonuclease protection assays and other applications where a low molecular weight RNA ladder is useful. One tube contain 5 μg (0.5mg/mL).
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Conditions: Room Temperature
Specifications
RNA Marker | |
Denaturing Polyacrylamide Gels | |
0.1 to 1 kb | |
5 μg | |
Nucleic Acid Gel Electrophoresis & Blotting |
Store at –20°C. | |
Room Temperature | |
Unlabled | |
Ambion™ | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.