missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Blaubrand™ IntraEND™ Disposable Micropipets

Comprised of disposable glass without ring mark. BRAND™ Blaubrand™ IntraEND™ Disposable Micropipets are in compliance with DIN ISO 7550 and CE-marked in accordance with IVD-Directive 98/79. Volume is defined by end-to-end filling. Volumes greater than 5μL include DE-M marking.

Brand:  Brand™ 709101

UNSPSC : 41121709

Code : 03

Additional Details : Weight : 0.01000kg

Product Code. 10634331

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Comprised of disposable glass without ring mark.
  • Complies with DIN ISO 7550.
  • CE Marked in accordance with IVD-Directive 98/79
  • Calibrated to contain (TC, In.)
  • Volume is defined by end-to-end filling
  • Volumes greater than 5μL include DE-M marking.
Specifications

Specifications

≤±0.5%
DIN ISO 7550, IVD-Directive 98/79 CE
32mm
1μL
'To Contain'
Yes
250/Box
Glass