missing translation for 'onlineSavingsMsg'
Learn More

Brand™ intraEND BLAUBRAND™ Disposable Delbrück MicroPipet

Brand:  Brand™ 708664

Code : 03

Additional Details : Weight : 0.07000kg

Product Code. 10429451

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

≤±0.5%
30mm
50μL
Yes
1000/Pack
Videos