missing translation for 'onlineSavingsMsg'
Learn More

Interference Check Standard 5 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 984

Code : 04

Additional Details : Weight : 0.59000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 500mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 11425908

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
Ca6000μg/mL
Fe5000μg/mL
Mg3000μg/mL
Al1200μg/mL
Na1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

Interference Check Standard 5
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specifications

Interference Check Standard 5
For Environmental Analysis
Triple-leached LDPE bottle
5% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
7697-37-2
Components in 5% HNO3:
Al    •1200μg/mL
Ca    •6000μg/mL
Fe    •5000μg/mL
Mg    •3000μg/mL
Na    •1000μg/mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
Videos

Product intended for laboratory use only