missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Interchangeable Block for PCMT Thermoshaker

Catalog Number 12337627
753.34 GBP valid until 2024-12-31
Use promo code "21906" to get your promotional price.


Click to view available options
No. of Places:
1 96-well Microplate
24
32
Holds:
0.5mL and 1.5mL microtubes
1.5mL microtubes
1.5mL microtubes plus 2.0mL microtubes
2.0mL microtubes
96-well microplate (0.2mL)
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides excellent uniformity and mixes evenly across the thermoshaker block. Grant Instruments™ Interchangeable Block for PCMT Thermoshaker is quick to reach specified mixing speed.

  • A block is required for use with PCMT Thermoshaker. Specify when ordering
  • Choice of 5 interchangeable blocks including a block suitable for microplates:
    • HC18 20 (20 x 0.5 ml and 12 x 1.5 ml microtubes)
    • HC32 (20 x 0.2 ml and 12 x 1.5 ml microtubes)
    • HC24N (24 x 1.5 ml microtubes)
    • HC24 (24 x 2 ml microtubes)
    • HC96 (96-well microplate for PCR, 0.2 ml) 
Holds 0.5mL and 1.5mL microtubes
No. of Places 32
Temperature Range (Metric) 4°C to 100°C
Type Block
Temperature Uniformity ±0.1°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'