missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7, FITC, clone: FIB504, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5867-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516576

  • £62.63 / 25 tests


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The FIB504 monoclonal antibody reacts with human and mouse integrin beta 7. Integrin beta 7 is a 130 kD glycoprotein which associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Main ligands for integrin alpha 4 beta 7 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the main ligand of integrin alpha E beta 7 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response. The FIB504 antibody has been reported to have blocking activity.Applications Reported: This FIB504 antibody has been reported for use in flow cytometric analysis.Applications Tested: This FIB504 antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Integrin Beta 7
FIB504
FITC
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin b7, ITGB7
IgG2a
RUO
Primary
Murine
Videos