missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 5, clone: KN52, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0497-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15287007

  • £87.86 / 100µg


EU Stock 2
Add to basket

Description

Description

Description: The KN52 antibody reacts with human integrin beta 5, an approximately 100,000 Mr (reduced) and 95,000 Mr (nonreduced) a member of the integrin family. Only a single alpha chain, the alpha v subunit, associates with the integrin beta 5 to form the vitronectin receptor complex. Integrin alpha v/beta 5 complex also binds to the basic domain of Tat (he sequence RKKRRQRRR). The integrin beta 5 is found on carcinoma cell lines, hepatoma and fibroblast cell lines, and is absent from lymphoblastoid cells and platelets.The KN52 monoclonal antibody has been shown to inhibit migration.Applications Reported: This KN52 antibody has been reported for use in flow cytometric analysis.Applications Tested: This KN52 antibody has been tested by flow cytometric analysis of Chinese Hamster Ovary (CHO), A549 and normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Integrin Beta 5
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
O70309, P18084
CD61, GP3A, GPIIIa
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HamsterþHumanþMurine
Flow Cytometry
KN52
Unconjugated
ITGB5
Purified
ITGB5
IgG1, kappa
100μg
Store at 2-8°C.
Primary
3693
Videos

For Research Use Only.