missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 3 Mouse anti-Human, Clone: 23C6, R&D Systems™
Mouse Monoclonal Antibody has been used in 13 publications
POA
Specifications
Antigen | Integrin alpha V beta 3 |
---|---|
Clone | 23C6 |
Immunogen | Human osteoclasts |
Gene ID (Entrez) | 3685 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF-ready.Specifications
Integrin alpha V beta 3 | |
Human osteoclasts | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Integrin alpha V beta 3. | |
Unconjugated | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
23C6 | |
3685 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
For Research Use Only