missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 4/CD49d Rat anti-Mouse, Clone: 265329, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Integrin alpha 4/CD49d |
---|---|
Clone | 265329 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 4/CD49d Tyr34-Phe976 Accession # Q792F9 |
Gene ID (Entrez) | 3676 |
Gene Accession No. | Q792F9 |
Description
Integrin alpha 4/CD49d Monoclonal antibody MAB2450SP Specifically detects Integrin alpha 4/CD49d in Mouse samples. It is validated for Flow CytometrySpecifications
Integrin alpha 4/CD49d | |
Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 4/CD49d Tyr34-Phe976 Accession # Q792F9 | |
Q792F9 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
265329 | |
3676 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse Integrin alpha 4/CD49d. | |
Unconjugated | |
Flow Cytometry, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only