missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4 beta 7 (LPAM-1), Biotin, clone: DATK32 (DATK-32), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5887-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15249899

  • £178.31 / 500µg


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

Description: The DATK32 monoclonal antibody binds to a combinatorial epitope of mouse LPAM-1 (alpha4beta7), which is a heterodimer of the the 154kDa alpha4 subunit and the 130kDa beta7 subunit. In mouse, the beta7 subunit is found on the majority of mature lymphocytes and a small population of thymocytes and bone marrow cells. LPAM-1 is an integrin involved in the transendothelial migration of lymphocytes across high endothelial venules (HEV), and is know to bind several ligands including MAdCAM-1, VCAM-1 and fibronectin. Binding of the DATK32 monoclonal antibody has been shown to induce aggregation of the CD8+ T cell lymphoma TK1, and block LPAM-1-mediated cell adhesion.Applications Reported: This DATK32 (DATK-32) antibody has been reported for use in flow cytometric analysis.Applications Tested: This DATK32 (DATK-32) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Integrin alpha 4 beta 7 (LPAM-1)
DATK32 (DATK-32)
Biotin
Conjugated
IgG2a, kappa
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
500μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos

For Research Use Only.