missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4 beta 7 (LPAM-1), APC, clone: DATK32 (DATK-32), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5887-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15350770

  • £62.64 / 25µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DATK32 monoclonal antibody binds to a combinatorial epitope of mouse LPAM-1 (alpha 4/ beta 7), which is a heterodimer of the the 154kDa alpha 4 subunit and the 130kDa beta 7 subunit. In mouse, the beta 7 subunit is found on the majority of mature lymphocytes and a small population of thymocytes and bone marrow cells. LPAM-1 is an integrin involved in the transendothelial migration of lymphocytes across high endothelial venules (HEV), and is know to bind several ligands including MAdCAM-1, VCAM-1 and fibronectin. Binding of the DATK32 monoclonal antibody has been shown to induce aggregation of the CD8+ T cell lymphoma TK1, and block LPAM-1-mediated cell adhesion.Applications Reported: This DATK32 (DATK-32) antibody has been reported for use in flow cytometric analysis.Applications Tested: This DATK32 (DATK-32) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Integrin alpha 4 beta 7 (LPAM-1)
DATK32 (DATK-32)
APC
Conjugated
IgG2a
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos