Learn More
Integrated Gas Module for Varioskan™ LUX
Control O2 and CO2 levels with this optional, integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader.
Brand:  Thermo Scientific VLUPG006
Code : NEW
Additional Details : Weight : 1.20000kg
 View more versions of this product
Product Code. 15320787
  • £10208.88 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Disclaimers:
NOTE: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications
Specifications
Integrated gas module upgrade kit (service upgradable)
0.1 to 15%
1 to 21%
Varioskan LUX
± 0.3% at 5% CO2
±0.3% at 1% O2
Videos