missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Insulysin/IDE Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | Insulysin/IDE |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Insulysin/IDE Polyclonal specifically detects Insulysin/IDE in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Insulysin/IDE | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, IDE, INSDEGM, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
IDE | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P14735 | |
3416 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE Met42-Leu1019 Accession # P14735 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Insulysin/IDE in direct ELISAs and Western blots. |
For Research Use Only