missing translation for 'onlineSavingsMsg'
Learn More

Insulin Mouse, eFluor 570, Clone: ICBTACLS, eBioscience™

Mouse Monoclonal Antibody

£73.93 - £170.59

Specifications

Antigen Insulin
Clone ICBTACLS
Host Species Murine
Applications Immunohistochemistry, Microscopy
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15568966
View Documents
Affymetrix eBioscience
41-9769-80
25μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15578966
View Documents
Affymetrix eBioscience
41-9769-82
100μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This ICBTACLS monoclonal antibody reacts with the hormone insulin, responsible for regulation of carbohydrate and fat metabolism. Blood glucose levels are maintained by insulin. Insulin secretion from pancreatic beta-cells within the islets of Langerhans results in uptake of excess glucose from the blood into organs and its storage in the form of glycogen. Patients with type 1 diabetes have an autoimmune destruction of insulin-producing beta-cells in the pancreas, resulting in insulin deficiency and elevated levels of blood glucose. Patients with type 2 diabetes have insulin resistance where insulin becomes less effective at reducing the amount of blood glucose. Insulin is also important for proper endothelial cell function in with respect to vascular elasticity in response to changing blood pressure and plays a role in cognition, learning, and memory.

The ICBTACLS antibody recognizes human, mouse, and rat insulin.
Specifications

Specifications

Insulin
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ICBTACLS
Immunohistochemistry, Microscopy
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL

For Research Use Only.