missing translation for 'onlineSavingsMsg'
Learn More

Insulin, clone: ICBTACLS, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9769-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537666

  • £172.54 / 100µg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This ICBTACLS monoclonal antibody reacts with the hormone insulin, responsible for regulation of carbohydrate and fat metabolism. Blood glucose levels are maintained by insulin. Insulin secretion from pancreatic beta-cells within the islets of Langerhans results in uptake of excess glucose from the blood into organs and its storage in the form of glycogen. Patients with type 1 diabetes have an autoimmune destruction of insulin-producing beta-cells in the pancreas, resulting in insulin deficiency and elevated levels of blood glucose. Patients with type 2 diabetes have insulin resistance where insulin becomes less effective at reducing the amount of blood glucose. Insulin is also important for proper endothelial cell function in with respect to vascular elasticity in response to changing blood pressure and plays a role in cognition, learning, and memory.The ICBTACLS antibody recognizes human, mouse, and rat insulin.Applications Reported: This ICBTACLS antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy.Applications Tested: This ICBTACLS antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low or high pH antigen retrieval and can be used at less than or equal to 0.25 μg/mL. This ICBTACLS antibody has been tested by western blot of non-reduced pancreatic tissue lysate and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Insulin
ICBTACLS
Unconjugated
Purified
IgG2a
RUO
Primary
Rat
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C.
Monoclonal
Videos