Learn More
MP Biomedicals™ Insulin, Human Recombinant
Insulin, Human Recombinan, ∽28 IU/mg, Powder
Brand: MP Biomedicals™ 0219390080
186.23 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 11061-68-0 Weight : 0.00010kg
Description
- Methods for immobilizing insulin on polystyrene dishes have been reported for applications involving protein-free cell cultures.
- Insulin is the primary polypeptide hormone responsible for controlling the cellular uptake, utilization and storage of glucose, amino acids and fatty acids while inhibiting the breakdown of glycogen, protein and fat.
- It is produced by pancreatic beta cells.
Specifications
5807.6 | |
Store at -20°C. | |
Powder |
100mg | |
Proteins, Enzymes & Peptides |