Learn More
Thermo Scientific Chemicals Insulin B (22-25)
For research use
Brand: Thermo Scientific™ J61170.MC
Code : NEW
Additional Details : CAS Number : 34367-73-2 Weight : 0.00010kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lyophilized powder | |
-30° to -10°C | |
H-Arg-gly-Phe-Phe-OH | |
Native |
100mg | |
RUO | |
Insulin B (22-25) | |
Lyophilized |
RUO – Research Use Only