missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standard 8 For Environmental Analysis, SPEX CeriPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 662

Code : 04

Additional Details : Weight : 0.59000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 500mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 11415898

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
Al50μg/mL
As50μg/mL
Co50μg/mL
Cr50μg/mL
Cu50μg/mL
K50μg/mL
Na50μg/mL
P50μg/mL
Pb50μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

ICP Instrument Check Standard 8
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specifications

ICP Instrument Check Standard 8
Each component at 50μg/mL in 2% HNO3:
Aluminum
Arsenic
Chromium
Cobalt
Copper
Lead
Phosphorus
Potassium
Sodium
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
For Environmental Analysis
Triple-leached LDPE bottle
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Videos

Product intended for laboratory use only